Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laccaic Acid from Lacca, TCI America™
SDP

Catalog No. L009525G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

60687-93-6

Molecular Formula

C91H62N2O44

Molecular Weight (g/mol)

1887.467

MDL Number

MFCD00071249

InChI Key

NGZUCVGMNQGGNA-UHFFFAOYSA-N

PubChem CID

133109320

IUPAC Name

7-[5-(2-acetamidoethyl)-2-hydroxyphenyl]-3,5,6,8-tetrahydroxy-9,10-dioxoanthracene-1,2-dicarboxylic acid;7-[5-(2-amino-2-carboxyethyl)-2-hydroxyphenyl]-3,5,6,8-tetrahydroxy-9,10-dioxoanthracene-1,2-dicarboxylic acid;3,5,6,8-tetrahydroxy-7-[2-hydroxy-5-(2-

SMILES

CC1=C2C(=CC(=C1C(=O)O)O)C(=O)C3=CC(=CC(=C3C2=O)O)O.CC(=O)NCCC1=CC(=C(C=C1)O)C2=C(C3=C(C(=C2O)O)C(=O)C4=CC(=C(C(=C4C3=O)C(=O)O)C(=O)O)O)O.C1=CC(=C(C=C1CCO)C2=C(C3=C(C(=C2O)O)C(=O)C4=CC(=C(C(=C4C3=O)C(=O)O)C(=O)O)O)O)O.C1=CC(=C(C=C1CC(C(=O)O)N)C2=C(C3=C(C(=

Specifications

Quantity

25 g

Physical Form

Crystalline Powder

Chemical Name or Material

Laccaic Acid from Lacca

RTECSNumber

GD2425100

TSCA

No

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.