Learn More
Ted Pella Inc LACEY FORMVAR/CARBON 300M 50/P
SDP
Supplier:  Ted Pella Inc 01883F
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Grid, Lacey Formvar/Carbon, 300 Mesh, Copper, Grid Hole Size: ~63microm, A Lacey Formvar film coated with a heavy layer of carbon
Catalog No. NC9157220