Learn More
Ted Pella Inc LACEY FORMVAR S W/CARBON 200M
SDP
Supplier:  Ted Pella Inc 01881F
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Grid, Lacey Formvar/Carbon, 200 Mesh, Copper, Grid Hole Size: ~ 97microm, A Lacey Formvar film coated with a heavy layer of carbon, 50/PK
Catalog No. NC9114960
missing translation for 'QuantityLeft'