Learn More
LaCrosse™ Premium Five-Buckle Overshoes
Made of a premium rubber compound with high tensile strength for durability and fit over leather work boots
$207.50
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-130-2624
|
LaCrosse™
26714015 |
15 |
Pair of 1 for $207.50
|
|
|||||
Description
- Ozone-resistant, abrasion-resistant rubber offers 100% waterproof protection
- 3M™ Scotchlite reflective material provides visibility in low-light conditions
- Designed to fit over deep heel footwear
- Heavy-duty, industrial strength buckles allow for easy on and off
- ZTX rubber maintains flexibility in cold conditions
- Non-corrosive metal buckles offer an adjustable fit
- Abrasion-resistant Hex-Tread outsole provides superior traction
- Black overshoes with a height of 35.56cm (14 in.)
Specifications
14 in. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.