Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Lactose, Monohydrate
SCI_ED

Catalog No. p-4416324 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 64044-51-5
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.312
InChI Key WSVLPVUVIUVCRA-ZEAJSSQNSA-N
Synonym d-lactose monohydrate, d-+-lactosemonohydrate
PubChem CID 133126686
IUPAC Name (3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O

Specifications

Color White
Quantity 100 g
Assay Percent Range 100% w/v
Linear Formula C12H22O11·H2O
Formula Weight 360.32
Grade Laboratory
Packaging Lab Grade
Density 1.525g/mL
Chemical Name or Material Lactose, Monohydrate
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.