Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina Biological™ Lady Beetle 10-Larvae Culture
SDPSCI_ED

Catalog No. S72460 Shop All Carolina Biological Supply Products
Click to view available options
:

Includes

10-15 Coleomegilla maculata larvae, food and care instructions.

Observe all 4 stages of complete metamorphosis with lady beetle larvae.

  • Alligator-like larvae grow, pupate and metamorphose in 10 to 14 days.
  • At least 5 of the 10 to 15 larvae guaranteed to emerge as healthy adult lady beetles.

Note: Prepaid coupons allow you to order your organisms in advance. We'll send a coupon(s) that you later redeem by phone, fax, Online web form, or mail. To ensure prompt arrival of material, coupons must be redeemed at least 2 weeks prior to requested delivery date.

Specifications

Content And Storage

22 to 25°C

Species

Lady Beetle

For Use With (Application)

Carolina lady bug larvae are an excellent alternative to butterflies in examining insect metamorphosis

Product Type

Larvae Culture

Quantity

10 to 15

Source

USA

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.