Learn More
PIP™ Lady Eva Reader Bifocal
PIP™ Lady Eva Reader Bifocal features a construction that better fits female facial structure
Supplier:  PIP™ 250120150
 View more versions of this product
Catalog No. 19-148-257
missing translation for 'QuantityLeft'
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Designed for the worker who needs to see up close
  • Offered in increments of +0.25 diopters to give users the exact reading power of their personal glasses
  • Lightweight, rimless design provides all day user comfort
  • Rubber co-molded temples provide a secure fit
  • Molded nose bridge design to fit the vast majority of users
  • 10 Base curve lens
  • Meets ANSI Z87.1+
Specifications
Specifications
Clear
+1.50
Polycarbonate
Clear, Pink
Polycarbonate
0.9999
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Title
Select an issue
Warning - Prop 65 - Reproductive Harm