Learn More
Revvity Health Sciences Inc LANCE Ultra ULight-phospho-40S ribosomal protein S6 (pSer235/236) Peptide, 5 nmoles
Supplier:  Revvity Health Sciences Inc TRF0129M
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
LANCE Ultra TR-FRET technology provides a two-component kinase assay that combines direct labeling of the kinase substrate with ULight, an innovative emission dye for optimum binding efficiency, reducing steps and assay prep time. Validated for over 310 kinases, we provide data transparency by supplying: Test conditions: Rapid assay setup to move straight into optimized assay performance S:B data: Confidence in proven performance of the kinase on specific substrates; no guessing Phospho-motifs: Substrate sequences are provided Assay options: Multiple substrates are tested so you can select the best peptide sequence and S:B for your kinase For ULight substrates: 0.5 nmoles 1,000 assay points 5 nmoles 10,000 assay points In LANCE Ultra kinase assays, the binding of a Eu-labeled anti-phosphosubstrate antibody to the phosphorylated ULight-labeled substrate brings donor and acceptor molecules into close proximity. After irradiation of the kinase reaction at 320 or 340 nm, the energy fro
Catalog No. 50-209-8673