Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
SDP

Catalog No. p-7131912 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 168106-26-1
Molecular Formula C6F18LaN3O12S6
Molecular Weight (g/mol) 979.312
InChI Key XQHMUWQUZHVCJH-UHFFFAOYSA-N
Synonym Bis(trifluoromethanesulfonyl)imide Lanthanum(III) Salt, Lanthanum(III) Triflimide
PubChem CID 129827648
IUPAC Name bis(trifluoromethylsulfonyl)azanide;lanthanum(3+)
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[La+3]

Specifications

Color White
Quantity 1 g
UN Number 1759
Formula Weight 979.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lanthanum(III) Bis(trifluoromethanesulfonyl)imide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.