Learn More
AdipoGen Lasalocid A Sol acetonitrile
SDP
Supplier:  AdipoGen CDXL0515LL25
Encompass_Preferred
Chemical. CAS 25999-20-6. Formula C34H53NaO8. MW 612.8. Synthetic. Ionophore. Potent antibacterial agent and a coccidiostat. Able to form neutral complexes with monovalent and divalent cations and transport them through apolar phase including lipid bilayer membranes. It can transport also larger organic cations, like protonated dopamine.
Catalog No. 50-256-0595
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.