Learn More
PIG™ Latching Drum Lid
SDP
Latching drum lid to keep drums and hazardous waste storage containers securely closed and locked.
Supplier:  PIG™ DRM659RD
 View more versions of this product
Catalog No. 18-366-518
Add to Cart
Description
Description
  • For 55 gal. Steel Drums
  • Bolt-Ring
  • Latching design opens and closes with one hand for easy in-and-out drum access - keep drums covered and avoid chemical waste fines
  • Nitrile gasket and locking mechanism seal lid to help reduce vapor emissions and keep drum contents dry and secure
  • Easy-access lid closes securely to help you meet hazardous waste regulations
  • Hinge stops keep lid upright when open
  • Durable, chemical-resistant powder coat helps ensure long service life
  • Lockable latch helps prevent unauthorized access
  • Hinge pin is made for easy lid removal without loosening band around drum
  • Multiple lid color options allow you to segregate drums for safe, compliant filling
Specifications
Specifications
Latching Drum Lid
Non-sterile
68 cm
59 cm
11 cm
Red
26.75 in.
23.25 in.
4.375 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.