Learn More
NEOGEN Lauryl Tryptose (LST) Broth, 500 g
Supplier: NEOGEN 700003057
Lauryl Tryptose (LST) Broth is used for the detection of coliform bacteria in water and wastewater. Lauryl Tryptose Broth is not intended for use in the diagnosis of disease or other conditions in humans.Lauryl Tryptose Broth is a selective medium for the detection of coliforms in water, dairy products and other foods. The American Public Health Authority (APHA) recommend Lauryl Tryptose Broth for the Most Probable Number Presumptive Test of coliforms in waters, effluent or sewage and as a confirmation test of lactose fermentation with gas production from milk samples and for the detection of coliforms in foods.Lauryl Tryptose Broth is prepared according to the formulation of Mallmann and Darby. Mallmann and Darby showed that tryptose at a concentration of 2% increased the early logarithmic growth phase when compared to meat peptone.For more information visit https://www.neogen.com/solutions/Microbiology/lauryl-tryptose-broth
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.