Learn More
Research Products International Corp LB Agar, Low Salt Formula, Granulated, 10 KG

Supplier: Research Products International Corp L2403310000.0

For propagation and maintenance of E. Coli. Dissolve 32 grams in 1 Liter distilled water and adjust the pH to 7.2. Sterilize by autoclaving for 15 minutes.
Ingredients:
Sodium Chloride: 5 g/L
Yeast Extract: 5 g/L
Agar: 12 g/L
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.