Learn More
Optimize Technologies All-In-One EXP Direct-Connect / In-Line Holder components (Fittings Included, for use w/EXP Trap and Guard Cartridges)
SDP
Supplier:  Optimize Technologies 150204041
Encompass_Preferred
The patent-pending Hand-Tight EXP™ Trap Column is rated for use at 20,000+ psi (1,400+ bar). This unique design connects directly to any injection valve (with 10-32 threads) or in-line with 1/16” stainless tubing for unparalleled convenience and efficiency. The EXP™ Cartridge System enables chemists to quickly remove detergents or salts which can affect the ionization process in MS work. This trapping technique can concentrate the sample directly on-line, allowing for increased recovery of precious sample material compared to off-line techniques. On-line trapping readily lends itself to automation for high-throughput analysis in UHPLC/MS applications. Free-Turn™ architecture allows the user to change cartridges by hand without breaking fluid connections on the holder inlet/outlet. This product includes both In-Line and Direct-Connect Holder components. Use with EXP™ Trap and Guard cartridges.
Catalog No. 50-753-3521
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.