Learn More
Optimize Technologies EXP In-Line Holder (Fittings Included, for use w/EXP Trap and Guard Cartridges)
SDP
Supplier:  Optimize Technologies 150203946
Encompass_Preferred
The EXP™ In-Line Holder is rated to 20,000 psi with hand-tight replaceable cartridges. This unique design connects in-line to any UHPLC system or injection valve with 1/16” stainless tubing and included EXP™ fittings. The EXP™ Cartridge System enables chemists to quickly remove detergents or salts which can affect the ionization process in MS work. This trapping technique can concentrate the sample directly on-line, allowing for increased recovery of precious sample material compared to off-line techniques. On-line trapping readily lends itself to automation for high-throughput analysis in UHPLC/MS applications. Free-Turn™ architecture allows the user to change cartridges by hand without breaking fluid connections on the holder inlet/outlet. This product features hardened stainless steel end caps eliminating galling, Optimize's famous Free-Turn architecture and comes packaged with Fittings. Use with EXP™ Trap and Guard Cartridges.
Catalog No. 50-753-3518
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.