Learn More
Eurofins Discoverx LCK Inhibitor, 1 mg. (92-1322)
Supplier:  Eurofins Discoverx 921322
Encompass
LCK (lymphocyte-specific protein tyrosine kinase) inhibitor is a pyrrolopyrimidine that blocks the activity of two forms of Lck kinase, Lck (64-509) and Lckcd, and has been shown to inhibit T cell receptor-stimulated IL-2 production. The LCK protein is a member of the Src-family of non-receptor protein tyrosine kinases and plays a critical role in the initial steps of T cell receptor signaling that trigger the production of cytokines. Provided as 1 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9835
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.