Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Acetate, Trihydrate, Crystal, Reagent, ACS, 99-103%, Spectrum™ Chemical
SDP

Catalog No. p-7157656 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

L1040, 6080-56-4, C4H6O4Pb•3H2O

Spectrum™ Chemical Lead Acetate, Trihydrate, Crystal, Reagent, ACS is a chemical compound and is obtained by treating lead oxide with acetic acid. It is used as a reagent and comes in a crystalline form. As an ACS grade Reagent, Spectrum Chemical manufactured compounds are used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 6080-56-4
Molecular Formula C4H12O7Pb
Molecular Weight (g/mol) 379.30
MDL Number MFCD00150023
InChI Key MCEUZMYFCCOOQO-UHFFFAOYSA-L
IUPAC Name λ²-lead(2+) diacetate trihydrate
SMILES O.O.O.[Pb++].CC([O-])=O.CC([O-])=O

Specifications

CAS Max % 100%
Quantity 125 g
UN Number UN1616
Percent Purity 99 to 103%
Grade ACS
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.