Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lead(II) niobium(V) 2-ethylhexanoate decaethoxide, 99% (metals basis), 10% w/v in ethanol, Thermo Scientific™
Click to view available options
Quantity:
25mL
100mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
UN Number | UN1170 |
Quantity | 25mL |
Sensitivity | Moisture Sensitive |
Formula Weight | 1130.04 |
Physical Form | Liquid |
Percent Purity | 99% |
Assay | (metals basis) |
Chemical Name or Material | Lead(II) niobium(V) 2-ethylhexanoate decaethoxide |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement H225-H311-H371-H332-H315-H319 Highly flammable liquid and vapor. Toxic in contact with skin. May cause damage to organs. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a Keep away from heat/sparks/open flames/hot surfaces. No smoking. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Remove/Take off immediately all contaminated clothing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction