Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) Phthalocyanine 95.0+%, TCI America™
SDP

Catalog No. p-7134758 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
25 g

Chemical Identifiers

CAS 15187-16-3
Molecular Formula C32H16N8Pb
MDL Number MFCD00053951
Synonym Phthalocyanine Lead(II)

Specifications

Color Blue-Purple
Quantity 1 g
UN Number 3077
Formula Weight 719.74
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lead(II) Phthalocyanine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.