Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Lead Nitrate, Crystal
SCI_ED

Catalog No. p-4564868 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
2.5 kg
Packaging:
Reagent Grade - Crystals

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 10099-74-8
Molecular Formula N2O6Pb
Molecular Weight (g/mol) 331.20
MDL Number MFCD00011153
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
Synonym lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
PubChem CID 24924
ChEBI CHEBI:37187
IUPAC Name λ²-lead(2+) dinitrate
SMILES [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color White
Quantity 100 g
Assay Percent Range >99%
Linear Formula Pb(NO3)2
Formula Weight 331.2
Packaging Reagent Grade - Crystals
Density 4.53g/mL
Chemical Name or Material Lead Nitrate
ShelfLife 5 years
DOTInformation DOT Class 5.1(6.1), II, : Oxidizer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.