Learn More
TARGETMOL CHEMICALS INC LECOZOTAN HCL 5MG
Supplier:  TARGETMOL CHEMICALS INC T27806
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Lecozotan HCl (SRA-333) is a potent and selective 5-HT antagonist that significantly enhances KCl-stimulated glutamate and acetylcholine release from the hippocampal dentate gyrus and has cognitive enhancing properties. Chronic administration of Lecozotan HCl did not induce 5-HT(1A) receptor tolerance or desensitization in a behavioral model demonstrating 5-HT(1A) receptor function. purity: 99%
Catalog No. 50-258-0574
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.