Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Leibovitz's L-15 Medium, GlutaMAX™ Supplement
SDP

Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue. L-15 is buffered by phosphates and free base amino acids instead of sodium bicarbonate.

$65.50

Specifications

Concentration 1 X
Form Liquid
Product Type Leibovitz's L-15 Medium
Without Additives No HEPES, No Sodium Bicarbonate, No Glucose
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
31-415-029
SDP
View Documents
Gibco™
31415029
500 mL
Each of 1 for $65.50
Add to Cart
 
Description

Description

Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue. L-15 is buffered by phosphates and free base amino acids instead of sodium bicarbonate. This medium is designed for supporting cell growth in environments without CO2 equilibration.

This L-15 is modified as follows:
With: Galactose, Phenol Red, GlutaMAX™, Sodium Pyruvate
Without: Glucose, HEPES, Sodium Bicarbonate

The complete formulation is available.

Gibco™ L-15 with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.

L-15 contains no proteins, lipids, or growth factors. Therefore, L-15 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS).

Specifications

Specifications

1 X
Leibovitz's L-15 Medium
cGMP-compliant under the ISO 13485 standard
Liquid
No HEPES, No Sodium Bicarbonate, No Glucose
7.4 to 7.9
Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 Months

SDS
Documents

Documents

Product Certifications

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.