Learn More
Cole-Parmer Shimpo DT-107A-S12 Digital Contact Tachometer, LED Display, 12" Standard Wheel
Supplier:  Cole-Parmer 9880737
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Eliminate inaccurate measurements, thanks to this rugged tachometer made of die-cast aluminum. Features direct length measurement, extensive speed range (0.10 to 25,000 rpm) and a large 5-digit display. Lengthy battery life.
Catalog No. 50-992-091