Learn More
Bioworld Lens culinaris (Lentil) Lectin (LCA/LCH) - FITC
Supplier:  Bioworld 217610332
Encompass_Preferred
Technical Specifications:Ligand Source:Lens culinarisMolecular Weight: 50 kDaCarbohydrate Specificity:αMan (High Mannose), Fucα6GlcNAcInhibiting/Eluting Sugar:a-Methylglucoside +a-MethylmannosideCarbohydrate for Elution: 0.2Ma-Methylglucoside +a-MethylmannosideBlood Group Specificity: N/AConjugate: FITCForm: LiquidIsoelectric (pI): 7.6-8.4Storage: -20°CApplication note: Calcium and Manganese are required for binding.
Catalog No. 50-199-146
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.