Learn More
TARGETMOL CHEMICALS INC LENSIPRAZINE 5MG
Supplier: TARGETMOL CHEMICALS INC T32633
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Lensiprazine (SLV314) is a potent dopamine D(2) receptor antagonist that acts as a serotonin reuptake inhibitor and can be used to study bipolar disorder and schizophrenia. purity: 98%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.