Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Letrozole, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, reversible non-steroidal aromatase inhibitor (IC50 = 11.5 nM). Displays antitumor effects in several animal models. Suppresses the endogenous aromatase-induced proliferation of MCF-7 cells.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 285.3 |
Percent Purity | >99% |
Chemical Name or Material | Letrozole |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction