Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leuco Crystal Violet 98.0+%, TCI America™
SDP

Catalog No. p-7132112 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
250 g

Chemical Identifiers

CAS 603-48-5
Molecular Formula C25H31N3
Molecular Weight (g/mol) 373.54
MDL Number MFCD00008314
InChI Key OAZWDJGLIYNYMU-UHFFFAOYSA-N
Synonym leucocrystal violet, leuco crystal violet, leucomethyl green, leucogentian violet, 4,4',4-methylidynetris n,n-dimethylaniline, c.i. basic violet 3, leuco, crystal violet leucobase, tris p-dimethylaminophenyl methane, benzenamine, 4,4',4-methylidynetris n,n-dimethyl, 4,4',4-hexamethyltriamino triphenylmethane
PubChem CID 69048
IUPAC Name 4-{bis[4-(dimethylamino)phenyl]methyl}-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)C(C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 178°C
Color Green-White
Quantity 25 g
Formula Weight 373.54
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Leuco Crystal Violet
EINECSNumber (4)-1476
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.