Learn More
Selleck Chemical LLC Levetiracetam UCB-L059-50mg
Levetiracetam (UCB-L059, SIB-S1) is an anticonvulsant medication used to treat epilepsy. Levetiracetam (UCB-L059, SIB-S1) is an agonist of muscarinic acetylcholine receptors (mAChR). Levetiracetam modulates the presynaptic P/Q-type voltage-dependent calcium (Ca2+) channel.