Learn More
TARGETMOL CHEMICALS INC LIAFENSINE 5MG
Supplier:  TARGETMOL CHEMICALS INC T32737
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Liafensine(BMS-820836) is a novel and selective triple monoamine reuptake inhibitor with inhibitory effects on the reuptake of serotonin norepinephrine and dopamine for the study of major depressive disorder and central nervous system disorders. purity: 99%
Catalog No. 50-258-0323
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.