Learn More
LifeScan™ SureStep™ System
Designed for personal or bedside use
Specifications
For Use With (Equipment) | Sure step system |
---|
Includes: Kit Contents:
- SureStep™
- Meter
- 25 SureStep™
- Test Strips
- PENLET™
- II Automatic Blood Sampler
- 25 Lancets
- Logbook
- Two AAA Batteries
- Carrying Case
- Control Solution
- Owner's Manual†
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
23-029305
|
LifeScan™
53885048303 |
Surestep Pro Controls 3/pk | Surestep Pro Controls 3/pk | N/A | N/A | N/A | |||
23-029306
|
LifeScan™
53885063702 |
Surestep Control Solutn 2vl/pk | Surestep Control Solutn 2vl/pk | N/A | N/A | N/A | |||
Description
- Stores up to 150 test results
- Results in 15 seconds
- Glucose range: 0 to 500mg/dL; hematocrit range: Adult 25 to 60%
- Offers testing date/time stamp and QC event flag
- Extra-large display
- Ten-test memory
- Blue confirmation dot indicates adequate sample application
† Includes SureStep™ Meter, operating guide and quick reference manual
Specifications
Sure step system |