Learn More
Electron Microscopy Sciences Retriever Thermal Slide Processor Lifting Device
Supplier: Electron Microscopy Sciences 6270503
Used with the Retriever benchtop thermal slide processer that is used on formalin-fixed, paraffin embedded tissues prior to immunostaining.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.