Learn More
Cole-Parmer Extech 407026 High-Intensity, Dual-Scale Light Meter
Supplier:  Cole-Parmer 0158832
Encompass_Preferred
Three-range meter indicates light intensity up to 50,000 lux or 5000 footcandles for five selectable lighting types. The RS-232 interface and optional 4000-reading datalogger module facilitate data documentation (sold separately).
Catalog No. 50-992-893
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.