Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Limonin, >99.6%, For HPLC analysis, MP Biomedicals™

Catalog No. p-4405624 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
5 mg
10 mg
25mg
50mg

Limonin from Citrus Seeds White Crystalline Powder

  • It may be used in assays to quantify limonin in citrus products.
  • It may also be used in other assays as a control, standard, or competitor or in studies on the physiology, metabolism, and biosynthesis of limonin.
  • It is a natural compound found in grapefruit and other citrus seeds.
  • IT is a triterpenoid aglycone that is a bitter principle of citrus fruits.
  • It has been shown to inhibit chemically induced carcinogenesis and demonstrates anti feedant properties.

Chemical Identifiers

CAS 1180-71-8
Molecular Formula C26H30O8
Molecular Weight (g/mol) 470.52
MDL Number MFCD00075922
InChI Key KBDSLGBFQAGHBE-MSGMIQHVSA-N
PubChem CID 179651
ChEBI CHEBI:16226
IUPAC Name (1R,2R,7S,10R,13R,14R,16S,19S,20S)-19-(furan-3-yl)-9,9,13,20-tetramethyl-4,8,15,18-tetraoxahexacyclo[11.9.0.0²,⁷.0²,¹⁰.0¹⁴,¹⁶.0¹⁴,²⁰]docosane-5,12,17-trione
SMILES CC1(C)O[C@H]2CC(=O)OC[C@@]22[C@H]1CC(=O)[C@]1(C)[C@@H]2CC[C@@]2(C)[C@@H](OC(=O)[C@H]3O[C@@]123)C1=COC=C1

Specifications

Color White
Assay Percent Range >99.6%
Quantity 5 mg
Synonym limonin, dictamnolactone, obaculactone, citrolimonin, evodin, 7,16-dioxo-7,16-dideoxylimondiol, limoni, limonoic acid, di-delta-lactone, limonine, limonoate d-ring-lactone
Formula Weight 470.5
Physical Form Powder
Chemical Name or Material Limonin, For HPLC analysis
Recommended Storage Store at 2° to 8°C.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.