Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AUDIT™ MicroControls™ Linearity FD Cardiac Markers for Ortho Vitros
SDP

Catalog No. 22158628
Click to view available options
:
This item is not returnable. View return policy
This item is not returnable. View return policy

Linearity FD Cardiac Markers for Ortho Vitros is assayed quality control material consisting of five levels of human serum.

  • Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
  • Free and easy to use Auditor™ QC “instant reports” data reduction software or AUDIT™ Faxback.

Specifications

Content And Storage

2°C to 8°C

Detectable Analytes

Creatinine Kinase-MB (CK-MB), Myoglobin, Troponin I

For Use With (Equipment)

Ortho Vitros

Sample Type

Whole Blood, Serum, Plasma

Type

Linearity

For Use With (Application)

It is intended to simulate human patient serum samples for purpose of determining linearity, calibration verification and verification of reportable range for Anti-SARSCoV-2 S.

Level

5 (A-E)

Quantity

5 x 1 mL

Stability

3 Days

Form

Freeze-dried

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.