Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AUDIT MicroControls Linearity FD Procalcitonin for Siemens Atellica
SDP

Catalog No. 22158642
Click to view available options
:
This item is not returnable. View return policy
This item is not returnable. View return policy

Assayed quality control material consisting of five levels of bovine based serum containing the analyte: Procalcitonin.

  • Assayed quality control material
  • Five levels of bovine based serum albumin
  • Analyte: Procalcitonin

Specifications

Certifications/Compliance

CLIA

Content And Storage

Store at 2°C to 8°C

Concentration

1 mL

Detectable Analytes

Procalcitonin (PCT)

For Use With (Equipment)

Siemens Atellica Instruments

Sample Type

Whole Blood, Serum, Plasma

Type

Vial

Disposable

Single-use

For Use With (Application)

Quantitative Assays on the Indicated Analyzer provided in the labeling. The Linearity FD Procalcitonin for Siemens Atellica should not be used for calibration or standardization of the Procalcitonin assays. The Linearity FD Procalcitonin for Siemens Atellica is for In Vitro Diagnostic use only.

Includes

5 levels of 1 mL bovine based serum containing the following analyte: Procalcitonin.

Intended Use

It is intended to simulate human patient serum samples for purpose of determining linearity, calibration verification, and verification of reportable range for Procalcitonin.

Level

1, 2, 3, 4, 5

Quantity

5/Cs.

Source

Bovine-based serum

Stability

When used to monitor the precision of laboratory testing procedures for its assays, Linearity FD Procalcitonin for Siemens Atellica has a reconstituted stability of up to 5 days under the proper storage conditions. Leaving the vial uncapped, or prolonging its time (at room temperature, will void this open vial stability claim. Make sure the contents of the vial are well mixed before use.

Form

Freeze-dried

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.