Learn More
Beckman Coulter Beckman Coulter LINERS SYSTEM, 1L, QTY 6
Supplier:  Beckman Coulter C15216
Encompass_Preferred
Beckman Coulter HarvestLine system disposable non-sterile liners, 1L, used with the 1L bottles for the JLA-9.1000 and JLA-8.1000 fixed angle rotors. (Qty 6)
Catalog No. 50-163-2004
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.