Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Linopirdine dihydrochloride, Tocris Bioscience™

Catalog No. p-4784109 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

KV7 (KCNQ) channel blocker

Blocker of KV7 (KCNQ) voltage-gated potassium channels; blocks KV7.1 + 7.3 (KCNQ2 + 3)/ M-currents (IC50 = 4 - 7 μM) and KV7.1 (KCNQ1) homomeric channels (IC50 = 8.9 μM). Augments hippocampal ACh release and is a cognitive enhancer following oral administration in vivo.

Chemical Identifiers

CAS 113168-57-3
Molecular Formula C26H23Cl2N3O
Molecular Weight (g/mol) 464.39
InChI Key ZEVVHCGTTNRYOY-UHFFFAOYSA-N
Synonym linopirdine dihydrochloride, 1-phenyl-3,3-bis pyridin-4-ylmethyl indolin-2-one dihydrochloride, c26h21n3o.2hcl, 1,3-dihydro-1-phenyl-3,3-bis 4-pyridinylmethyl-2h-indol-2-one dihydrochloride, 3,3-bis 4-pyridylmethyl-1-phenylindolin-2-one dihydrochloride
PubChem CID 14209557
IUPAC Name 1-phenyl-3,3-bis(pyridin-4-ylmethyl)indol-2-one;dihydrochloride
SMILES C1=CC=C(C=C1)N2C3=CC=CC=C3C(C2=O)(CC4=CC=NC=C4)CC5=CC=NC=C5.Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 464.39
Percent Purity >99%
Chemical Name or Material Linopirdine dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.