Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium bis(trimethylsilyl)amide, Thermo Scientific Chemicals

Catalog No. p-7052122
Click to view available options
Quantity:
5 g
25 g

Base used in preparation of dienes and enolates

Lithium bis(trimethylsilyl)amide is a base used in preparation of dienes and enolates. It is used to catalyze the addition of phosphine P-H bonds to carbodiimides leading to phosphaguanidines. Lithium bis(trimethylsilyl)amide is also used in a novel three-step synthesis of disubstituted 1,2,5-thiadiazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon

Chemical Identifiers

CAS 4039-32-1
Molecular Formula C6H18LiNSi2
Molecular Weight (g/mol) 167.327
MDL Number MFCD00008261
InChI Key YNESATAKKCNGOF-UHFFFAOYSA-N
Synonym lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide
PubChem CID 2733832
IUPAC Name lithium;bis(trimethylsilyl)azanide
SMILES [Li+].C[Si](C)(C)[N-][Si](C)(C)C

Specifications

Melting Point 83°C to 86°C
Color White
Linear Formula [(CH3)3Si]2NLi
Quantity 5 g
UN Number UN2925
Beilstein 8136861
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 167.33
Physical Form Powder
Chemical Name or Material Lithium bis (trimethylsilyl)amide
Show More Show Less
Hazard Category H228-H314
Hazard Statement GHS H Statement
H228-H314-H318
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 223-725-6
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.