Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium cyclopentadienide, 97%, Thermo Scientific Chemicals

Catalog No. p-7050716
Click to view available options
Quantity:
5 g
25 g

Lithium cyclopentadienide is a useful reagent in preparing metallocenes and cyclopentadienyl derivatives. It is also used as intermediates and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16733-97-4
Molecular Formula C5H5Li
Molecular Weight (g/mol) 72.04
MDL Number MFCD00001439,MFCD00001439
InChI Key CSOHRTAOCDVTQU-UHFFFAOYSA-N
Synonym lithium cyclopentadienide, cyclopentadienyllithium, acmc-1bo1f, lithium cyclopenta-1,3-dien-1-ide, lithium 1+ ion cyclopenta-1,3-dien-1-ide
PubChem CID 14063469
IUPAC Name lithium;cyclopenta-1,3-diene
SMILES [Li+].C1C=CC=[C-]1

Specifications

Quantity 5 g
UN Number UN3180
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 72.04
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Lithium cyclopentadienide
Hazard Category H314
Hazard Statement GHS H Statement
H228-H314-H318
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.