Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Locust (Carob) Bean Gum, FCC, Spectrum™ Chemical
SDP

Catalog No. 18603962 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
Packaging:

L1135, 9000-40-2

Spectrum™ Chemical Locust (Carob) Bean Gum, FCC is a gelling and thickening agent used in certain foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS

9000-40-2

Specifications

CAS Max %

100%

Loss on Drying

14%

Packaging

Fiber Drum

Quantity

50 kg

Grade

FCC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.