Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Loreclezole hydrochloride, Tocris Bioscience™

Catalog No. p-4783602 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Subtype-selective GABAA modulator

Subtype-selective GABAA receptor modulator. Acts as a positive allosteric modulator of β2 or β3-subunit containing receptors. Also acts as a negative modulator at a novel regulatory site, enhancing GABAA receptor sensitization. Inhibits homomeric ρ1 GABAC receptors.

Chemical Identifiers

CAS 2227372-56-5
Molecular Formula C10H7Cl4N3
Molecular Weight (g/mol) 310.987
InChI Key AWKSVFLKVCVVFQ-MDZFRNKHSA-N
Synonym loreclezole hydrochloride, dsstox_cid_25676, dsstox_rid_81050, dsstox_gsid_45676, cas-nocas_45676, z-1-2-chloro-2-2,4-dichlorophenyl vinyl-1h-1,2,4-triazole hydrochloride, z-1-2-chloro-2-2,4-dichlorophenyl ethenyl-1h-1,2,4-triazole hydrochloride
PubChem CID 45073437
IUPAC Name 1-[(Z)-2-chloro-2-(2,4-dichlorophenyl)ethenyl]-1,2,4-triazole;hydrochloride
SMILES C1=CC(=C(C=C1Cl)Cl)C(=CN2C=NC=N2)Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 311
Percent Purity >99%
Chemical Name or Material Loreclezole hydrochloride
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.