Learn More
Apexbio Technology LLC Losartan Potassium (DuP 753),1g CAS# 124750-99-8
Selective non-peptide angiotensin AT1 receptor antagonist. Inhibits the contractile effects of angiotensin II on rabbit aorta and jugular vein (pA2 = 8.27). Orally active antihypertensive agent. Other sizes are also available. Please inqury us for quote.