Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Loxoprofen Sodium Salt Dihydrate 98.0+%, TCI America™
SDP

Catalog No. p-7131893 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 226721-96-6
Molecular Formula C15H21NaO5
Molecular Weight (g/mol) 304.318
MDL Number MFCD01745788
InChI Key BAZQYVYVKYOAGO-UHFFFAOYSA-M
Synonym Sodium 2-[4-[(2-Oxocyclopentyl)methyl]phenyl]propionate, 2-[4-[(2-Oxocyclopentyl)methyl]phenyl]propionic Acid Sodium Salt
PubChem CID 23674745
ChEBI CHEBI:31786
IUPAC Name sodium;2-[4-[(2-oxocyclopentyl)methyl]phenyl]propanoate;dihydrate
SMILES CC(C1=CC=C(C=C1)CC2CCCC2=O)C(=O)[O-].O.O.[Na+]

Specifications

Melting Point 198°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 268.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Loxoprofen Sodium Salt Dihydrate
RTECSNumber CY1678845
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.