Learn More
Electron Microscopy Sciences LR Gold Resin 500 ML
Supplier:  Electron Microscopy Sciences 14371
Encompass_Preferred
The London Resin Company's acrylic "LR Gold" offers the histochemist and immunocytochemist the advantage of working with unfixed tissues at low temperatures, thus allowing many fixation-sensitive procedures to be carried out in 1-4 microns resin sections at -25 degree C using visible light. It is non-toxic and has a low viscosity.
Catalog No. 50-980-418
missing translation for 'QuantityLeft'
Explore available promotions
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.