Learn More
AdipoGen Lucigenin
SDP
Supplier:  AdipoGen CDXD0068G010
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS 2315-97-1. Formula C28H22N4O6. MW 510.5. Synthetic. Chemiluminescent probe for the detection of peroxides in biological systems. Reagent used for the generation of chemiluminescence in the presence of a reductant and oxygen It is employed in numerous chemiluminescent assays. Lucigenin is specific for superoxide anion radical.
Catalog No. 50-256-1265
missing translation for 'QuantityLeft'