Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LUF 5834, Tocris Bioscience™

Catalog No. p-4835944 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent adenosine A2A and A2B receptor partial agonist

LUF 5834 is a potent A2A and A2B adenosine receptor partial agonist (Ki = 2.6 nM and EC50 = 12 nM respectively). Also exhibits selectivity for A1 over A3 (Ki values are 2.6 and 538 nM respectively).

Chemical Identifiers

CAS 333962-91-7
Molecular Formula C17H12N6OS
Molecular Weight (g/mol) 348.384
InChI Key LOLJUQOCQBYBGT-UHFFFAOYSA-N
Synonym 2-1h-imidazol-2-yl methyl thio-6-amino-4-4-hydroxyphenyl pyridine-3,5-dicarbonitrile, 2-amino-4-4-hydroxyphenyl-6-1h-imidazol-2-ylmethyl thio-3,5-pyridinecarbonitrile, 2-amino-4-4-hydroxy-phenyl-6-1h-imidazol-2-ylmethylsulfanyl-pyridine-3,5-dicarbonitrile
PubChem CID 10066330
IUPAC Name 2-amino-6-(1H-imidazol-2-ylmethylsulfanyl)-4-(4-oxocyclohexa-2,5-dien-1-ylidene)-1H-pyridine-3,5-dicarbonitrile
SMILES C1=CC(=O)C=CC1=C2C(=C(NC(=C2C#N)SCC3=NC=CN3)N)C#N

Specifications

Quantity 10 mg
Formula Weight 348.38
Chemical Name or Material LUF 5834
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.