Learn More
New England Biolabs, Inc. LunaScript Multiplex One-Step RT-PCR Kit - 250 reactions
The LunaScript Multiplex One-Step RT-PCR Kit offers a streamlined protocol for cDNA synthesis and PCR amplification in a single reaction. It contains a reaction mix and an enzyme mix featuring Luna WarmStart Reverse Transcriptase and Q5 Hot Start High Fidelity DNA polymerase. The kit is optimized for multiplex target amplification to enable diagnostics pathogen detection and viral genome sequencing.