Learn More
TARGETMOL CHEMICALS INC LURASIDONE 100MG
Supplier:  TARGETMOL CHEMICALS INC T21337
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg and bulk. Please contact Fisher for quotes. Lurasidone (SM-13496) is an antagonist of both dopamine D2 and 5-HT7(IC50=1.68 and 0.495 nM respectively).It also a partial agonist of 5-HT1A receptor(IC50 6.75 nM). purity: 99%
Catalog No. 50-258-1029
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.