Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Luria Agar (Miller's) Tubes
SCI_ED

Catalog No. S802486 Shop All Aldon Products
Click to view available options
:
Luria Agar Tubes; Media Type: Luria Agar; Format: 12 x 6 mL Tubes; Type: Agar Slants; Form: Solid
This item is not returnable. View return policy
This item is not returnable. View return policy

Luria Agar (Miller's) Tubes are manufactured sterile microbiological media for the lab and education market. The classic black rubber lined phenolic caps ensure a sterile seal. Great quality and competitive pricing will help you outsource or upgrade your current product line.

  • Phenolic caps ensure sterile seal
  • Great quality
  • Competitive pricing

Specifications

Detects Bacterial Growth
Form Solid
Format 6 mL Tubes
Grade High School
Manufacturing Origin United States
Media Type Luria Agar
Preparation Method Autoclave
Product Type Agar Slants
Sterility Sterile
Target Organism Class Fastidious and Non-fastidious Organisms
For Use With (Application) Growing Bacteria
No. per Pack 12 Pk.
Packaging Type Box
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.