Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LY 266097 hydrochloride, Tocris Bioscience™

Catalog No. p-4816839 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective 5-HT2B receptor antagonist

Selective 5-HT2B receptor antagonist (pKi = 9.3). More than 100-fold selective over 5-HT2A and 5-HT2C. Attenuates amphetamine-induced locomotion in the rat.

Chemical Identifiers

CAS 172895-39-5
Molecular Formula C21H24Cl2N2O2
Molecular Weight (g/mol) 407.335
InChI Key KPXKZZURYAXZQE-UHFFFAOYSA-N
Synonym ly 266097 hydrochloride, 1-2-chloro-3,4-dimethoxyphenyl methyl-2,3,4,9-tetrahydro-6-methyl-1h-pyrido 3,4-b indole hydrochloride, ly hplc , powder, 1-2-chloro-3,4-dimethoxybenzyl-6-methyl-1,2,3,4-tetrahydro-9h-pyrido 3,4-b indole hydrochloride, 1-2-chloro-3,4-dimethoxyphenyl methyl-6-methyl-1h,2h,3h,4h,9h-pyrido 3,4-b indole hydrochloride
PubChem CID 9953184
IUPAC Name 1-[(2-chloro-3,4-dimethoxyphenyl)methyl]-6-methyl-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole;hydrochloride
SMILES CC1=CC2=C(C=C1)NC3=C2CCNC3CC4=C(C(=C(C=C4)OC)OC)Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 407.33
Percent Purity >99%
Chemical Name or Material LY 266097 hydrochloride
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.